Refractometers
Medline Scientific™ CETI 4.5kg Convex Refractometer
Highly versatile refractometer that allows measurements of a wide range of substances. Medline Scientific™ CETI 4.5kg Convex Refractometer is capable of reading both reflected and transmitted light.
ATAGO™ RX-9000α Automatic Digital Refractometer
Designed with a wide measurement range, measurement temperature range, and high measurement accuracy, the ATAGO™ RX-9000α Automatic Digital Refractometer is suitable for multiple sample types such as measuring oils and fats with high melting points, aroma oils of high refractive index and organic solvents, meeting demands for high accuracy measurements.
ATAGO™ Digital Hand-Held Pocket Refractometer: PAL-03S
Designed for sodium chloride measurement (g/100g). The Atago PAL-03S Digital Hand-Held Pocket Refractometer provides accuracy of ±0.2%.
ATAGO™ RX-5000α-Bev Automatic Digital Refractometer
Ideal for measuring beverages. The ATAGO™ RX-5000α-Bev Automatic Digital Refractometer is a redesigned version of the conventional Rx-5000μ. A flat sample stage replaced the sample well to make it easier to wipe off the sample and to allow for a faster and easier clean up. The RX-5000μ-Bev is ideal for measuring beverages and is equipped with all the standard features of the RX-5000μ.
ATAGO™ NAR-3T Abbe Refractometer
The ATAGO™ NAR-3T Abbe Refractometer was developed to give improved measuring accuracy and ease of use. This has been achieved by making fundamental improvements to the optical system, utilizing a larger scale, incorporating a high intensity lamp and using a double control knob to give quick and finer control.
ATAGO™ RX-5000α Automatic Digital Refractometer
Can set measurement temperature internally and measures the refractive index, Brix or concentration of various liquids accurately and speedily. The ATAGO™ RX-5000α Automatic Digital Refractometer has a thermo-module to control temperature, so a constant temperature water bath is not necessary.
ATAGO™ ATAGO™ PR-32α digitale brix-refractometer
Covers low-concentration Brix values of fruit juices, other food and beverage products as well as chemicals and industrial liquids. The ATAGO™ PR-32α Digital Brix Refractometer is an upgraded model of the PR-32 with improved measurement accuracy based on the new technology development.
ATAGO™ Digital Hand-Held Pocket Refractometer: PAL-3
Full range refractometer for measuring almost any sample. The ATAGO™ Digital Hand-Held Pocket Refractometer, PAL-3 has a measurement time of 3 seconds.
ATAGO™ ATAGO™ digitale draagbare zakrefractometer: PAL-1
Breed meetbereik. De Atago digitale draagbare zakrefractometer, PAL-1, is ideaal voor het meten van vrijwel alle fruitsappen, voedsel en dranken.
METTLER TOLEDO™ Refracto™ 30PX Portable Refractometer
The handheld digital refractometer returns accurate and precise results within seconds, displayed in the units of measurement needed. Designed for measurements in laboratory and production environments.
Hanna Instruments™ Digital Refractometers
Easy to use tool for measuring sugar in food samples in the field or in the lab that is ideal for the analysis of fruits, energy drinks, puddings, soy milk, juices, jam, marmalade, honey, soups, jelly, tofu and condiments.
METTLER TOLEDO™ Refracto™ 30GS Portable Refractometer
The handheld digital refractometer returns accurate and precise results within seconds, displayed in the units of measurement needed. Offers an extended measuring range due to its special prism design. Used for measurements in laboratory and production environments.
Bellingham + Stanley™ Eclipse Hand Held Refractometer
Manufactured to the highest specification using only the best materials and optical components. Bellingham + Stanley™ Eclipse Hand Held Refractometer is a low-cost, portable instrument used for on-the-spot determination of Refractive Index of liquids or the concentration of solutions.
Atago™ MASTER-53M Refractometer
Easy-to-read, easy-to-hold, and smooth-sampling refractometer. Atago™ MASTER-53M Refractometer is suitable for a wide range of solution sampling.
Atago™ HSR–500 Hand-Held Refractometer
Ideal for measuring sugar concentration in solution. Atago™ HSR–500 Hand-Held Refractometer measures the refractive index of a solution containing sugar, which is proportional to sugar concentration.
ATAGO™ Abbe Refractometer NAR-1T
The Abbe Refractometer NAR-1T SOLID is suitable to measure the Refractive Index Scale and the Brix scale for liquid and solid samples. Operates with D-line (589 nm) light source.
Reichert™ Refractometers voor industriële vloeistoftesten
Designed specifically for checking ratio or concentration of water-soluble industrial fluids