Smeltkroezen
Porcelaines Avignon™ Hoge smeltkroezen, porselein
High form, glazed porcelain crucible with a high resistance to thermal and physical shock. Porcelaines Avignon™ High Form Porcelain Crucibles are chemically neutral with an enamel that is unalterable to most concentrated acids.
Fisherbrand™ PTFE Crucibles
Guaranteed inert and crack-free. Fisherbrand™ PTFE Crucibles can be used up to 280°C.
Haldenwanger™ Aluminium keramische conische lage smeltkroes
Use these crucibles for general laboratory applications. Haldenwanger™ Aluminum Ceramic Conical Squat Form Crucibles are resistant to a variety of acids and alkalis.
Haldenwanger™ Lids for Porcelain Crucibles
Use these lids with porcelain crucibles. Haldenwanger™ Lids for Porcelain Crucibles are constructed from optimized laboratory porcelain.
Haldenwanger™ Hoge smeltkroezen zonder deksel, porselein
Use these dishes for a variety of applications. Haldenwanger™ Tall Form Porcelain Crucibles feature a smooth internal finish, constructed from optimized laboratory porcelain.
Haldenwanger™ Deksels voor porseleinen smeltkroezen
Designed to fit Haldenwanger™ Porcelain Crucibles.
Porcelaines Avignon™ Nacelle rechthoekig porseleinen mandje
Add the Porcelaines Avignon™ Nacelle Rectangular Porcelain Basket to your porcelain laboratory glassware for effective and efficient heating and cooling of mixtures.
Thermo Scientific™ Glassy Carbon E-Beam Evaporating Crucible
Glassy carbon offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids.
Thermo Scientific™ Graphite, Fusion Crucible, drillpoint, unpurified, volume 7.88cc
Manufactured from graphite materials that provide exceptional thermal uniformity and exactness to dimensional specifications.
DWK Life Sciences Kimble™ KIMAX™ Gooch Filtering Crucibles
Dry precipitates to constant weight at 110°C (230°F)
Heraeus™ High Form Platinum Crucible
Use for various fusion or melting processes in the analytical lab. Heraeus™ High Form Platinum Crucible is a range of containers with good resistance to aggressive media and stability even at high temperatures. Crucibles use a dispersion hardened allot for increased strength and service times.
Fisherbrand™ High-Form Porcelain Crucibles
Bestand tegen temperaturen tot 1150 °C (2102 °F)
Fisherbrand™ Porcelain Crucible Covers
For use with Fisherbrand High- and Wide-Form Porcelain Crucibles.