Ultrasoonapparaten, homogenisatoren en blenders
IKA Generatoren met rotor/stator voor IKA Ultra-Turrax™ homogenisatoren
Suitable for dispersing instrument T 18 digital
Fisherbrand™ Pre-Filled Bead Mill Tubes
For use with Bead Mill 4 and Bead Mill 24 homogenizers. May also be used with most other commercial Bead Mills.
IKA™ ULTRA-TURRAX™ T 18 Digital Disperser
Disperses volumes from 1 to 1500mL (H2O) with digital speed display. IKA™ ULTRA-TURRAX™ T 18 Digital Disperser offers a wide speed range from 3000 to 25,000 rpm that enables users to work at high circumferential speeds even with small rotor diameters. A broad choice of dispersing elements guarantees a wide range of applications.
Fisherbrand™ Q705 Sonicator with Probe
Use this high powered sonicator for any application and sample volume. Fisherbrand™ Sonicators are ideal for cell disruption, nanoparticle dispersion, DNA shearing/ChIP and homogenization.
Fisherbrand™ Bulk tubes
For use with Bead Mill 4 and Bead Mill 24 homogenizers. Also fits most other commercially available Bead Mills.
Kinematica Polytron™ 2500E Homogenizer
Versatile units combine quality, convenience and high performance for rapid, reproducible sample processing and consistent sample-to-sample uniformity
Fisherbrand™ Cup Horn for Model 505 and 705 Sonic Dismembrator
Cup Horn Accessory for use with Model 505/705 Sonic Dismembrator. Indirectly process multiple small samples (1ml or less) in sealed tubes. Prevent cross contamination.
Fisherbrand™ 505 Sonicator with Probe
Use this high powered sonicator for various applications and sample volumes. Fisherbrand™ Sonicators with Probe are ideal for cell disruption, nanoparticle dispersion, homogenisation and sample preparation.
Fisherbrand™ Krik
Ideal for raising samples up to a stationary probe mounted within the enclosure or on a stand.
Sonics Materials VCX-130PB Ultrasonic Processor
Robust and reliable 130 watt ultrasonic processor with thumb-actuated pulser for small volume applications.
Fisherbrand™ Sondes voor sonische dismembrators model 500 en 700
Microtips are thin, high intensity probes which are designed for processing small sample volumes. Microtips screw into the threaded end of the standard 1/2 in. probe (4220).
DWK Life Sciences Kimble™ Solid Borosilicate Glass Beads for Distillation Columns
Resist attack by liquids or vapors
Fisherbrand™ Temperature Probe Accessory for FB705 Sonicator
For monitoring temperature of sensitive samples
Fisherbrand™ Replacement Tips for Standard Sonic Dismembrator Probes, Models Q500 and Q700
Replace eroded tips easily
Fisherbrand™ 850 Homogenizer Accessory, Generator Probe
Stainless-steel generator probe; Fisher Scientific™ 850 Homogenizer Accessory, Generator Probe is compatible with Fisher Scientific 850 Homogenizers.
Thermo Scientific™ Wig-L-Bug™ Mixing/Grinding Mill Accessory, Ball Pestles, 9.52mm Dia.
For use with Wig-L-Bug Mixing/Grinding Mill (AA42627KT)
Thermo Scientific™ Wig-L-Bug™ Mixing/Grinding Mill Accessory, Agate Vial
For use with Wig-L-Bug Mixing/Grinding Mill (AA44204KT and AA44604KT)
Fisherbrand™ Power Cables for Model 505 and 705 Sonic Dismembrator
Power Cable with European plug for Model 505/705 Sonic Dismembrator
Fisherbrand™ Q500 Sonicator without Probe
Providing programmable operation and a digital display of operating parameters. Fisherbrand™ Q500 Sonicator without Probe is excellent for nanoparticle dispersion, creating emulsions, cell lysis and homogenization applications.
Thermo Scientific™ Ball pestle; clear plexiglas; 1/8in. diameter
For use with Wig-L-Bug Mixing/Grinding Mill (AA44204KT and AA44604KT)
Cole-Parmer™ Propellers for ServoDyne™ Electronic Mixers
Three-bladed propeller for general purpose mixing
Fisherbrand™ Replacement Converter for Model 505 and 705 Sonic Dismembrator
Replacement 20kHz Converter for Model 505/705 Sonic Dismembrator
Fisherbrand™ Geluiddempende kap voor sonische dismembrator, modellen 505/705
Vermindert het geluid van sonische dismembratoren aanzienlijk
Fisherbrand™ Sonische dismembrator, model 50
Economical, small volume cell disruptor.
Thermo Scientific™ Wig-L-Bug™ Mixing/Grinding Mill Accessory, Stainless-Steel Vial
For use with Wig-L-Bug Mixing/Grinding Mill (AA42627KT)
Fisherbrand™ 150 and 850 Hand Held Homogenizer Accessory, Plastic Disposable Generator Probe
Compatible with Fisher Scientific™ 150 and 850 Homogenizers. Fisher Scientific™ 150 and 850 Hand Held Homogenizer Accessory, Plastic Disposable Generator Probe is ideal for applications where cross contamination is a concern. Adapter required for use.
Fisherbrand™ Sonische dismembrator, model 120
Programable system ideal for small volume liquid processing. Includes 1/8 in. probe for processing 0.5-15ml volumes. There are 3 probe size options available which allow for processing 0.2-50mL volumes.
Fisherbrand™ Sondes voor sonische dismembrators model 50 en 120
Image shows (left to right): 4423, 4422, 4435.