Vochtmeters
Fisherbrand™ Plastic Baumé Hydrometer, 0-25 Bé°
For measuring the density of liquids heavier than water. Fisherbrand™ Plastic Baumé Hydrometer, 0-25 Bé° is a shatterproof hydrometer made of HACCP standard plastic suitable for use with food.
H-B Instrument™ Durac™ Specific Gravity/Relative Density (g/cm3) Plain Form Hydrometers: for Liquids Heavier Than Water
Available in a variety of models measuring specific gravity/relative density (g/cm3) of liquids heavier than water in various ranges.
Alla France™ Glazen hydrometer
Use these hydrometers for a variety of applications. Alla France™ Glass Hydrometers feature a benchmark engraved on the stem to show any displacement of the scale.
Fisherbrand™ ASTM™ Specific Gravity 275mL Hydrometers for Liquids Heavier than Water
Accuracy meets or exceeds ASTM E 100 specifications.
Fisherbrand™ Hydrometer met dubbele schaal
A dual scale, universal plain form hydrometer for measuring specific gravity/relative density (g/cm3) of liquids lighter than or heavier than water and Baume in various ranges.
Fisherbrand™ Nauwkeurige specifieke-zwaartekrachthydrometers, lengte 340 mm
Requires 275mL liquid volume for submersion.
Fisherbrand™ Plastic Hydrometer, 0-35% Brix
A saccharometer used for measuring the sugar content of a solution. Fisherbrand™ Plastic Hydrometer, 0-35% Brix is a shatterproof hydrometer made of HACCP proof plastic suitable for use with food.
Fisherbrand™ ASTM™ Specific Gravity Hydrometers for Liquids Lighter than Water
Accuracy of hydrometer meets or exceeds ASTM™ E 100 specifications.
Fisherbrand™ ASTM™ Specific Gravity 260mL Hydrometers for Liquids Heavier than Water
Accuracy meets or exceeds ASTM E 100 specifications.
Fisherbrand™ Precision Hydrometers for Specific Gravity
High-precision instruments for determination of density in fluids. Fisherbrand™ Precision Hydrometers for Specific Gravity do not include a thermometer in the body and feature a reference temperature of 60/60°F.
Fisherbrand™ Precision Baumé Hydrometer, 30-35 Bé°
For measuring the density of liquids heavier than water. Fisherbrand™ Precision Baumé Hydrometer, 30-35 Bé° is packed in a square plastic sleeve and is supplied without a thermometer.
Fisherbrand™ Traceable™ digitale hygrometer/thermometer
Ideaal voor het continu bewaken van laboratoria, opslagfaciliteiten en fabrieken. Fisher Scientific Traceable digitale hygrometer/thermometer toont vochtigheidsgraad en temperatuur in één oogopslag op een led van 0.5 inch (1.2 cm).
Fisherbrand™ Baume Hydrometers
Baume “heavy” scale for working with liquids heavier than water.
Fisherbrand™ Soil Analysis ASTM Hydrometers
These hydrometers are used to determine the specific gravity of soil in the range of 0.995 to 1.038 or -5 to 60g/L (using a weight by volume scale).
H-B Instrument™ Durac™ Safety Specific Gravity/Relative Density (g/cm3) Combined Form Thermo-Hydrometers
Offers the additional convenience of an enclosed thermometer with a temperature scale of 0° to 150°F
H-B Instrument™ Durac™ Specific Gravity/Relative Density Plastic Plain Form Hydrometers
Shatterproof polycarbonate prevents contamination
H-B Instrument™ Durac™ Isopropyl Alcohol Hydrometers
Measures the percent by volume of isopropyl alcohol
H-B Instrument™ Durac™ Safety Precision Combined Form Hydrometers, API
These combined-form hydrometers, with convenient, built-in thermometer, measure the relative density (specific gravity) of oil and petroleum related products in various ranges from -1° to 101°API
H-B Instrument™ Durac™ Salt Brine Plastic Plain Form Hydrometers
Lead free and shatterproof for ultimate safety
H-B Instrument™ Durac™ Specific Gravity Precision Hydrometers
Used in food and beverage, chemical, pharmacology, transportation, refrigeration industries
H-B Instrument™ Durac™ Specific Gravity/Relative Density (g/cm3) and Baume Plain Form Hydrometers: for Liquids Heavier Than Water
Dual scale, plain form hydrometers measure specific gravity/relative density (g/cm3) of liquids heavier than water, and Baume in various ranges
H-B Instrument™ DURAC™ Digital Stopwatches
Supplied with multi-language (English, French, German, Italian, Portuguese and Spanish) instructions, these electronic stopwatches include a calendar and a clock.
H-B Instrument™ Durac™ Specific Gravity ASTM Hydrometers: for Heavy Liquids
Available in a variety of models measuring specific gravity of heavy and light liquids in various ranges from 0.650 to 2.000
H-B Instrument™ Durac™ Specific Gravity / Relative Density (g/cm3) Precision Hydrometers; Traceable to NIST
Measure specific gravity or relative density (g/cm³) of a variety of liquids with these hydrometers. H-B Instrument Durac™ Specific Gravity / Relative Density (g/cm³) Precision Hydrometers are ideal for industries such as food and beverage, chemical, pharmacology, transportation and refrigeration.
H-B Instrument™ Durac™ Battery Hydrometer with Siphon Set
Used to determine the specific gravity/relative density (g/cm3) of battery acid in the range of 1.150 to 1.300
H-B Instrument™ Durac™ Specific Gravity/Relative Density (g/cm3) Plain Form Hydrometers: for Liquids Lighter Than Water
Available in a variety of models measuring specific gravity/relative density (g/cm3) of liquids lighter than water in various ranges from 0.600 to 1.000.
H-B Instrument™ Durac™ Salt Brine Plain Form Hydrometers
Choose from two plain form models with ranges from 0 to 100% or 0 to 26.5% to measure sodium chloride saturation of salt brines