Learn More
ATAGO™ PR-32α Digital Brix Refractometer
Covers low-concentration Brix values of fruit juices, other food and beverage products as well as chemicals and industrial liquids. The ATAGO™ PR-32α Digital Brix Refractometer is an upgraded model of the PR-32 with improved measurement accuracy based on the new technology development.
Brand:  ATAGO™ 3405
Product Code. 10001112
  • 1055.00 € / Each
Explore more special offers
Description
Description
  • Unit is designed to cover low concentration Brix values of fruit juices, other food and beverage products as well as chemicals and industrial liquids such as cutting oil, cleaning liquid, and antifreeze
  • Programmed with the user-scale function which allows users to input the coefficient into the formula: [Concentration=Brix X Coefficient ] to display the concentration of varying sample types
Specifications
Specifications
Refractometer
PR-32α
Min. Indication 0.1%
5°C to 40°C
0.0 to 32.0 Brix
± 0.1%
Brix
5°C to 40°C