Jassen, schorten en overige laboratoriumkleding
Logistik™ Lancer-Style laboratoriumjassen die beschermen tegen spattende chemicaliën
Provide the answer to full frontal protection with Logistik™ Lancer-Style Chemical Splash Laboratory Coats The wrap-over style Lancer front coat helps repel liquid chemicals and provides the wearer with an excellent level of protection since the use of chemicals in a laboratory can prove a constant hazard to standard style open neck coats.
Logistik Unicorp™ Faithful laboratoriumjassen polyester/ katoen, wit
Use Logistik Unicorp™ Faithful White Ladies' Fitted Lab Coats for general laboratory safety. It features two lower patch pockets, breast pocket, concealed stud-fastened front and stud-fastened cuffs
Mutexil™ Beschermende CPE-schoenhoezen
Choose from a range of choices and construction, from sticky, anti-slip, fluid -resistant, polypropylene and CPE variations with Mutexil™ Protective Overshoe Coverings.
DuPont™ Tyvek 200 Easysafe Hooded Coverall
Offering adequate Type 5 and 6 barrier protection and remains both air and moisture vapour permeable. DuPont™ Tyvek™ 200 Easysafe Hooded Coveralls are made from a new and unique optimized polyethylene nonwoven that is softer with a more open structure than Tyvek™ 500 and 600.
Mutexil™ Beschermende mouwen van polypropyleen
Use Mutexil™ Polypropylene Protection Sleeves for general purposes over sleeves when environmental safety and sterility requirements dictate.
Mutexil™ Visitor Protective Lab Coats
Use Mutexil™ Visitor Protective Lab Coats for visitors' safety and protection when visiting a facility that merits this required protective clothing.
Kimberly-Clark™ A7 P+™ laboratoriumjassen
Gebruik deze jassen in laboratoriumomgevingen met een lage kans op blootstelling aan chemicaliën en een basale behoefte aan het onder controle houden van besmetting. Kimberly-Clark™ A7 P+™-laboratoriumjassen hebben elastische manchetten en een duimlus voor volledige polsbedekking. De extra lange mouwen helpen de handschoen op zijn plaats te houden.
Logistik Unicorp™ Faithful laboratoriumjassen 100% katoen, wit
Protect those working in the lab with a Logistik Unicorp™ Faithful White Cotton Lab Coat. The white coat comes in four unisex sizes, is not CE marked and conforms to Howie report guidelines.
3M™ White Disposable Protective Coveralls, 4545 Series
Biedt bescherming volgens PPE-richtlijn categorie III, type 5/6. De beschermende coverall van de serie 4545 beschermt tegen radioactieve deeltjes en biologische stoffen en vermindert de opbouw van statische lading.
3M™ Beschermende coverall van polypropyleen
Biedt bescherming volgens PPE-richtlijn categorie III, type 5/6. De beschermende coveralls van de serie 4515 beschermen tegen gevaarlijk stof zoals asbestvezels.
3M™ White Disposable Visitor Lab Coats, Series 4400
Use for hygiene applications. The 3M Disposable Visitor Lab Coats series is made from lightweight polypropylene.
3M™ Beschermende coveralls van polypropyleen
Biedt bescherming volgens PPE-richtlijn categorie III, type 5/6. De beschermende coveralls van de serie 4515 beschermen tegen gevaarlijk stof zoals asbestvezels.
3M™ White Protective Coverall 4520
Protective Coveralls that provide protection against liquid splashes and dust.
3M™ White Protective Coverall 4510
Protective Coverall 4510 offers economic protection against infective agents, certain hazardous dusts, and limited liquid splashes. The PE laminate includes micropores that allow heat and moisture to evaporate from the skin and out through the suit offering a light, comfortable, protective solution.
3M™ Orange Disposable Protective Coverall 4515
Protective Coverall 4515 Series protect from hazardous dusts such as asbestos fibers.
3M™ SMS beschermende coverall
Gebruik voor niet-gevaarlijke toepassingen. De blauwe wegwerpcoveralls 4500-serie biedt Categorie I bescherming van polypropyleen weefsel.
3M™ White Disposable Protective Coverall 4540+
Met uitstekende dubbele-materiaaltechnologie en CE-categorie III, type 5/6-bescherming is de beschermende coverall 4540-serie ideaal voor toepassingen waar hoge niveaus van zowel bescherming als comfort gewenst zijn.
3M™ Disposable Protective Oversleeve 444
Provides basic barrier protection
3M™ Disposable Protective Coveralls
With a breathable back panel to improve air ventilation and reduce heat build-up. 3M™ Protective Coveralls are manufactured from a high quality laminated material.
3M™ Witte beschermende coveralls van polypropyleen op SMS-basis, categorie I
Gebruik voor niet-gevaarlijke toepassingen. De witte coveralls van de 4500-serie voor eenmalig gebruik bieden Categorie I-bescherming van polypropyleen weefsel.
Thermo Scientific™ Cryo Aprons
Protect your clothing with Thermo Scientific™ Cryo Aprons, available in small, medium and large sizes.
3M™ Disposable Protective Overshoe Covers 442
The 3M™ Disposable Overshoe Covers 442 are disposable shoe covers designed to provide basic barrier protection to help protect footwear against light liquid splashes and hazardous dusts.
3M™ Gray Protective Coverall 4570
Protective Coverall 4570 helps protect against a range of chemical jets and sprays, and has been certified to Type 3/4/5/6 levels of protection.
Fisherbrand™ Tyvek™ Cat III Coverall
Fisherbrand Tyvek™ Cat III Hooded Coveralls made from a soft polyethylene nonwoven fabric, offering a great balance of protection and comfort. Providing adequate Type 5 and 6 barrier protection, they remain both air and moisture vapour permeable.
3M™ Laboratoriumjas voor eenmalig gebruik
Offers a barrier against non-hazardous particles and liquid splashes. The 3M Disposable Protective Lab Coat is anti-static treated and can be used in light duty applications.
3M™ Disposable Protective Overshoe Cover 403
The 3M™ Disposable Overshoe covers 402 are disposable shoe covers designed to provide basic barrier protection to help protect footwear against light liquid splashes and hazardous dusts.