missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Unhooded Coveralls

Clean-processed to maximize cleanliness and have been sterilized by gamma-irradiation. DuPont™ Tyvek™ IsoClean™ Unhooded Coveralls are folded to aid aseptic donning and individually packed in an ISO Class 4 cleanroom.

273.98 €

Specifications

Product Type Cleanroom Coveralls
Material HDPE
Color White
Ankle Style Elastic
Cuff Style Elastic
View More Specs

Products 3
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15521975
View Documents
DuPont™
D13471957
Medium This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15501975
View Documents
DuPont™
D13471819
2X-Large This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15511975
View Documents
DuPont™
D13471846
3X-Large This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Delivers an ideal balance of protection, durability and comfort.
  • Made of high density polyethylene using a patented flash spinning process.
  • Breathable and exceptionally low linting.
  • Garments and accessories made of clean-processed and sterile Tyvek™ IsoClean™ are typically used in cleanrooms within the biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments.
  • Clean-processed and sterilised by gamma-irradiation to SAL of 10-6 (ISO 11137-1)
  • Full traceability on all sterilized apparel with certificates of sterility available
  • Suitable for use in GMP class A/B (ISO Class 5) clean rooms
  • This garment is not CE certified as chemical protective clothing
Specifications

Specifications

Cleanroom Coveralls
White
Elastic
Bound
ISO Class 5
Class 100 (ISO 5)
HDPE
Elastic
Front Zipper
Sterile
Slip-retardant shoe covers
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.