Learn More
DuPont™ Tyvek™ IsoClean™ Unhooded Coveralls
Product Code. 15521975
Click to view available options
Size:
Medium
2X-Large
3X-Large
Clean-processed to maximize cleanliness and have been sterilized by gamma-irradiation. DuPont™ Tyvek™ IsoClean™ Unhooded Coveralls are folded to aid aseptic donning and individually packed in an ISO Class 4 cleanroom.
  • Delivers an ideal balance of protection, durability and comfort.
  • Made of high density polyethylene using a patented flash spinning process.
  • Breathable and exceptionally low linting.
  • Garments and accessories made of clean-processed and sterile Tyvek™ IsoClean™ are typically used in cleanrooms within the biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments.
  • Clean-processed and sterilised by gamma-irradiation to SAL of 10-6 (ISO 11137-1)
  • Full traceability on all sterilized apparel with certificates of sterility available
  • Suitable for use in GMP class A/B (ISO Class 5) clean rooms
  • This garment is not CE certified as chemical protective clothing
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Coveralls
Size Medium
Material HDPE
Color White
Ankle Style Elastic
Cuff Style Elastic
Closure Type Front Zipper
Seam Type Bound
Sterility Sterile
Protection Class ISO Class 5
Description Slip-retardant shoe covers
Model D13471957
Cleanroom Class Class 100 (ISO 5)