Learn More
DuPont™ Tyvek™ IsoClean™ Unhooded Coveralls
Product Code. 15511975
Click to view available options
Size:
Medium
2X-Large
3X-Large
Clean-processed to maximize cleanliness and have been sterilized by gamma-irradiation. DuPont™ Tyvek™ IsoClean™ Unhooded Coveralls are folded to aid aseptic donning and individually packed in an ISO Class 4 cleanroom.
  • Delivers an ideal balance of protection, durability and comfort.
  • Made of high density polyethylene using a patented flash spinning process.
  • Breathable and exceptionally low linting.
  • Garments and accessories made of clean-processed and sterile Tyvek™ IsoClean™ are typically used in cleanrooms within the biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments.
  • Clean-processed and sterilised by gamma-irradiation to SAL of 10-6 (ISO 11137-1)
  • Full traceability on all sterilized apparel with certificates of sterility available
  • Suitable for use in GMP class A/B (ISO Class 5) clean rooms
  • This garment is not CE certified as chemical protective clothing
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Coveralls
Size 3X-Large
Material HDPE
Color White
Ankle Style Elastic
Cuff Style Elastic
Closure Type Front Zipper
Seam Type Bound
Sterility Sterile
Protection Class ISO Class 5
Description Slip-retardant shoe covers
Model D13471846
Cleanroom Class Class 100 (ISO 5)