missing translation for 'onlineSavingsMsg'
Learn More

ZNF746 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen ZNF746
Concentration 0.31 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16883015
View Documents
Proteintech
24543-1-AP-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16873015
View Documents
Proteintech
24543-1-AP-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The calculated molecular weight of ZNF746 is 69 kDa, but phosphorylated ZNF746 is about 80-85 kDa.

ZNF746 may be involved in transcriptional regulation.
Specifications

Specifications

ZNF746
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Human, Mouse
Q3U133, Q6NUN9
155061, 69228
ZNF746 Fusion Protein Ag21566
Primary
-20°C
ZNF746
0.31 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
zinc finger protein 746, ZNF746
Znf746
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.