Learn More
ZNF746 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24543-1-AP-150UL
 View more versions of this product
Product Code. 16873015
  • 410.00 € / 150µL
Explore more special offers
Description
Description
The calculated molecular weight of ZNF746 is 69 kDa, but phosphorylated ZNF746 is about 80-85 kDa.
ZNF746 may be involved in transcriptional regulation.
Specifications
Specifications
ZNF746
Polyclonal
Unconjugated
ZNF746
zinc finger protein 746, ZNF746
Rabbit
Antigen Affinity Chromatography
RUO
155061, 69228
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q3U133, Q6NUN9
Znf746
ZNF746 Fusion Protein Ag21566
150 μL
Primary
Human, Mouse