missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF746 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24543-1-AP-20UL
This item is not returnable.
View return policy
Description
The calculated molecular weight of ZNF746 is 69 kDa, but phosphorylated ZNF746 is about 80-85 kDa.
ZNF746 may be involved in transcriptional regulation.Specifications
ZNF746 | |
Polyclonal | |
Unconjugated | |
ZNF746 | |
zinc finger protein 746, ZNF746 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
155061, 69228 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3U133, Q6NUN9 | |
Znf746 | |
ZNF746 Fusion Protein Ag21566 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction