missing translation for 'onlineSavingsMsg'
Learn More

SLAIN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen SLAIN2
Concentration 0.31 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16832334
View Documents
Proteintech
13908-1-AP-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16822334
View Documents
Proteintech
13908-1-AP-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SLAIN2 is closely related to SLAIN1, a novel stem cell protein that is expressed at the stem cell and epiblast stages. Unlike SLAIN1, SLAIN2 is widely expressed and is thought to play a role in regulating microtubule (MT) growth and organization. SLAIN2 binds to end-binding proteins (EBs), cytoplasmic linker proteins (CLIPs) such as CLIP170, CLIP-associated proteins, and interacts with the MT polymerase ch-TOG. Depletion of SLAIN2 using RNAi led to disorganization of the radial MT array, indicating its importance in the regulation of MT growth and organization.
Specifications

Specifications

SLAIN2
Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
Q8CI08, Q9P270
305310, 57606, 75991
SLAIN2 Fusion Protein Ag4888
Primary
-20°C
SLAIN2
0.31 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
KIAA1458, SLAIN motif family, member 2, SLAIN2
SLAIN2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.