missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLAIN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13908-1-AP-150UL
This item is not returnable.
View return policy
Description
SLAIN2 is closely related to SLAIN1, a novel stem cell protein that is expressed at the stem cell and epiblast stages. Unlike SLAIN1, SLAIN2 is widely expressed and is thought to play a role in regulating microtubule (MT) growth and organization. SLAIN2 binds to end-binding proteins (EBs), cytoplasmic linker proteins (CLIPs) such as CLIP170, CLIP-associated proteins, and interacts with the MT polymerase ch-TOG. Depletion of SLAIN2 using RNAi led to disorganization of the radial MT array, indicating its importance in the regulation of MT growth and organization.Specifications
SLAIN2 | |
Polyclonal | |
Unconjugated | |
SLAIN2 | |
KIAA1458, SLAIN motif family, member 2, SLAIN2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
305310, 57606, 75991 | |
-20°C | |
Liquid |
Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8CI08, Q9P270 | |
SLAIN2 | |
SLAIN2 Fusion Protein Ag4888 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction