Learn More
SLAIN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13908-1-AP-20UL
 View more versions of this product
Product Code. 16832334
  • 145.00 € / 20µL
Explore more special offers
Description
Description
SLAIN2 is closely related to SLAIN1, a novel stem cell protein that is expressed at the stem cell and epiblast stages. Unlike SLAIN1, SLAIN2 is widely expressed and is thought to play a role in regulating microtubule (MT) growth and organization. SLAIN2 binds to end-binding proteins (EBs), cytoplasmic linker proteins (CLIPs) such as CLIP170, CLIP-associated proteins, and interacts with the MT polymerase ch-TOG. Depletion of SLAIN2 using RNAi led to disorganization of the radial MT array, indicating its importance in the regulation of MT growth and organization.
Specifications
Specifications
SLAIN2
Polyclonal
Unconjugated
SLAIN2
KIAA1458, SLAIN motif family, member 2, SLAIN2
Rabbit
Antigen Affinity Chromatography
RUO
305310, 57606, 75991
-20°C
Liquid
Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8CI08, Q9P270
SLAIN2
SLAIN2 Fusion Protein Ag4888
20 μL
Primary
Human, Mouse, Rat