missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPARGC1B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | PPARGC1B |
---|---|
Concentration | 0.31 mg/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16851905
|
Proteintech
22378-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16841905
|
Proteintech
22378-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PGC-1 beta is a tissue specific co-activator that enhances the activity of estrogen receptor alpha. The co-activation function relies on a bipartite transcriptional activation domain and two LXXLL motifs that interact with the AR2 domain of ER-alpha in an estrogen-dependent manner. Ectopic expression of this protein has been shown to lead to increased mitochondrial number and basal oxygen consumption.Specifications
PPARGC1B | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q86YN6 | |
133522, 170826 | |
PPARGC1B Fusion Protein Ag17566 | |
Primary | |
-20°C | |
PPARGC1B |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ERRL1, PERC, PGC 1 beta, PGC 1(beta), PGC1, PGC1B, PPAR gamma coactivator 1 beta, PPARGC 1 beta, PPARGC1, PPARGC1B | |
Ppargc1b | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |