missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPARGC1B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22378-1-AP-20UL
This item is not returnable.
View return policy
Description
PGC-1 beta is a tissue specific co-activator that enhances the activity of estrogen receptor alpha. The co-activation function relies on a bipartite transcriptional activation domain and two LXXLL motifs that interact with the AR2 domain of ER-alpha in an estrogen-dependent manner. Ectopic expression of this protein has been shown to lead to increased mitochondrial number and basal oxygen consumption.Specifications
PPARGC1B | |
Polyclonal | |
Unconjugated | |
PPARGC1B | |
ERRL1, PERC, PGC 1 beta, PGC 1(beta), PGC1, PGC1B, PPAR gamma coactivator 1 beta, PPARGC 1 beta, PPARGC1, PPARGC1B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
133522, 170826 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q86YN6 | |
Ppargc1b | |
PPARGC1B Fusion Protein Ag17566 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |