Learn More
PPARGC1B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22378-1-AP-150UL
 View more versions of this product
Product Code. 16841905
  • 410.00 € / 150µL
Explore more special offers
Description
Description
PGC-1 beta is a tissue specific co-activator that enhances the activity of estrogen receptor alpha. The co-activation function relies on a bipartite transcriptional activation domain and two LXXLL motifs that interact with the AR2 domain of ER-alpha in an estrogen-dependent manner. Ectopic expression of this protein has been shown to lead to increased mitochondrial number and basal oxygen consumption.
Specifications
Specifications
PPARGC1B
Polyclonal
Unconjugated
PPARGC1B
ERRL1, PERC, PGC 1 beta, PGC 1(beta), PGC1, PGC1B, PPAR gamma coactivator 1 beta, PPARGC 1 beta, PPARGC1, PPARGC1B
Rabbit
Antigen Affinity Chromatography
RUO
133522, 170826
-20°C
Liquid
Immunoprecipitation, Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q86YN6
Ppargc1b
PPARGC1B Fusion Protein Ag17566
150 μL
Primary
Human, Mouse