missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride Saturated Solution, Pure, Electrode Filling Solution, Fisher Chemical™
30.45 € - 76.50 €
Chemical Identifiers
CAS | 7447-40-7, 7732-18-5 |
---|---|
Molecular Formula | ClK |
Molecular Weight (g/mol) | 74.55 |
MDL Number | MFCD00011360 |
InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
PubChem CID | 4873 |
ChEBI | CHEBI:32588 |
SMILES | [Cl-].[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10744181
|
Fisher Chemical
J/2983/05 |
100 mL | Amber Glass Bottle | ||||||
15938776
|
Fisher Chemical
J/2983/07 |
250 mL | HDPE Plastic Bottle | ||||||
15948776
|
Fisher Chemical
J/2983/15 |
1 L | HDPE Plastic Bottle | ||||||
Description
Chemical Identifiers
7447-40-7, 7732-18-5 | |
74.55 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
CHEBI:32588 |
ClK | |
MFCD00011360 | |
4873 | |
[Cl-].[K+] |
Specifications
106°C | |
Liquid | |
MFCD00011360 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
potassium;chloride | |
4873 |
7447-40-7, 7732-18-5 | |
ClK | |
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
[Cl-].[K+] | |
74.55 | |
CHEBI:32588 |