missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Saturated Solution, Pure, Electrode Filling Solution, Fisher Chemical™

Product Code. 15938776
Click to view available options
Quantity:
1 L
100 mL
250 mL
Packaging:
Amber Glass Bottle
HDPE Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7447-40-7, 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Boiling Point 106°C
Physical Form Liquid
Quantity 250 mL
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite
IUPAC Name potassium chloride
Packaging HDPE Plastic Bottle
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'