missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Sulfate, Reagent grade, Honeywell
Reagent Grade, anhydrous,≥97%
83.35 € - 3235.26 €
Chemical Identifiers
CAS | 7487-88-9 |
---|---|
Molecular Formula | MgO4S |
Molecular Weight (g/mol) | 120.36 |
MDL Number | MFCD00011110 |
InChI Key | CSNNHWWHGAXBCP-UHFFFAOYSA-L |
Synonym | magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot |
PubChem CID | 24083 |
ChEBI | CHEBI:32599 |
SMILES | [Mg++].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15681820
|
Honeywell Research Chemicals
208094-500G |
500 g | N/A | ||||||
15671820
|
Honeywell Research Chemicals
208094-2KG |
2 kg | N/A | ||||||
15661820
|
Honeywell Research Chemicals
208094-2.5KG |
2.5 kg |
418.65 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15601830
|
Honeywell Research Chemicals
208094-5KG |
5 kg | N/A | ||||||
15691820
|
Honeywell Research Chemicals
208094-50KG |
50 kg | N/A | ||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
7487-88-9 | |
120.36 | |
CSNNHWWHGAXBCP-UHFFFAOYSA-L | |
24083 | |
[Mg++].[O-]S([O-])(=O)=O |
MgO4S | |
MFCD00011110 | |
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot | |
CHEBI:32599 |
Specifications
7487-88-9 | |
MgO4S | |
NONH for all modes of transport | |
CSNNHWWHGAXBCP-UHFFFAOYSA-L | |
magnesium;sulfate | |
24083 | |
120.37g/mol | |
Reagent | |
Vapor Density: <0.01 (vs air) |
Anhydrous | |
MFCD00011110 | |
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot | |
[Mg++].[O-]S([O-])(=O)=O | |
120.36 | |
CHEBI:32599 | |
≥97% | |
0.1mmHg (20°C) | |
Magnesium Sulfate |
Safety and Handling
missing translation for 'einecsNumber' : 231-298-2
missing translation for 'rtecsNumber' : OM4500000
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title