missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Sulfate, Reagent grade, Honeywell
Click to view available options
Quantity:
500 g
2 kg
2.5 kg
5 kg
50 kg
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Physical Form | Anhydrous |
Quantity | 2 kg |
UN Number | NONH for all modes of transport |
IUPAC Name | magnesium;sulfate |
Formula Weight | 120.37g/mol |
Percent Purity | ≥97% |
Grade | Reagent |
Vapor Pressure | 0.1mmHg (20°C) |
Density | Vapor Density: <0.01 (vs air) |
Chemical Name or Material | Magnesium Sulfate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'