missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate, Reagent grade, Honeywell

Product Code. 15691820
Click to view available options
Quantity:
500 g
2 kg
2.5 kg
5 kg
50 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, anhydrous,≥97%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7487-88-9
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00011110
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
PubChem CID 24083
ChEBI CHEBI:32599
SMILES [Mg++].[O-]S([O-])(=O)=O

Specifications

Physical Form Anhydrous
Quantity 50 kg
UN Number NONH for all modes of transport
IUPAC Name magnesium(2+) sulfate
Formula Weight 120.37g/mol
Percent Purity ≥97%
Grade Reagent
Vapor Pressure 0.1mmHg (20°C)
Density Vapor Density: <0.01 (vs air)
Chemical Name or Material Magnesium Sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.