missing translation for 'onlineSavingsMsg'
Learn More

LSM8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen LSM8
Concentration 0.3 mg/mL
Applications Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16886313
View Documents Promotion Details
Proteintech
10134-1-AP-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16876313
View Documents Promotion Details
Proteintech
10134-1-AP-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene encodes a member of the like-Sm family of proteins. The encoded protein consists of a closed barrel shape, made up of five anti-parallel beta strands and an alpha helix. This protein partners with six paralogs to form a heteroheptameric ring which transiently binds U6 small nuclear RNAs and is involved in the general maturation of RNA in the nucleus.
Specifications

Specifications

LSM8
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
Unconjugated
Rabbit
Mouse, Human, Rat
O95777, Q6ZWM4
296913, 51691, 76522
LSM8 Fusion Protein Ag0178
Primary
-20°C
LSM8
0.3 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
LSM8, NAA38, YJR022W
LSM8
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications