Learn More
LSM8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10134-1-AP-150UL
 View more versions of this product
Product Code. 16876313
  • 410.00 € / 150µL
Explore more special offers
Description
Description
This gene encodes a member of the like-Sm family of proteins. The encoded protein consists of a closed barrel shape, made up of five anti-parallel beta strands and an alpha helix. This protein partners with six paralogs to form a heteroheptameric ring which transiently binds U6 small nuclear RNAs and is involved in the general maturation of RNA in the nucleus.
Specifications
Specifications
LSM8
Polyclonal
Unconjugated
LSM8
LSM8, NAA38, YJR022W
Rabbit
Antigen Affinity Chromatography
RUO
296913, 51691, 76522
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O95777, Q6ZWM4
LSM8
LSM8 Fusion Protein Ag0178
150 μL
Primary
Human, Mouse, Rat