missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSM8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10134-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the like-Sm family of proteins. The encoded protein consists of a closed barrel shape, made up of five anti-parallel beta strands and an alpha helix. This protein partners with six paralogs to form a heteroheptameric ring which transiently binds U6 small nuclear RNAs and is involved in the general maturation of RNA in the nucleus.Specifications
LSM8 | |
Polyclonal | |
Unconjugated | |
LSM8 | |
LSM8, NAA38, YJR022W | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
296913, 51691, 76522 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95777, Q6ZWM4 | |
LSM8 | |
LSM8 Fusion Protein Ag0178 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |