missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptane Fraction, Honeywell Riedel-de Haën™
boiling range 93.5 - 100.4 °C
35.93 € - 122.19 €
Chemical Identifiers
CAS | 142-82-5 |
---|---|
Synonym | n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish |
PubChem CID | 8900 |
ChEBI | CHEBI:43098 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15685730
|
Honeywell-Riedel-de Haen
51760-1L |
1 L | Glass Bottle | ||||||
15695730
|
Honeywell-Riedel-de Haen
51760-2.5L |
2.5 L | Aluminum Bottle | ||||||
15605740
|
Honeywell-Riedel-de Haen
51760-5L |
5 L | Aluminum Bottle | ||||||
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Chemical Identifiers
142-82-5 | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
8900 | |
CHEBI:43098 |
Specifications
-90.5°C | |
C7H16 | |
UN1206 | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
CCCCCCC | |
100.21 | |
CHEBI:43098 | |
98.2°C to 98.4°C | |
1.397 |
142-82-5 | |
MFCD00009544 | |
1730763 | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
heptane | |
8900 | |
0.71g/mL at 20°C | |
−4°C (24.8°F) | |
Heptane Fraction |
Safety and Handling
P210-P261-P273-P301 + P310-P331-P501
missing translation for 'dotInformation' : class 3 - PG 2 - Heptanes