missing translation for 'onlineSavingsMsg'
Learn More

Heptane Fraction, Honeywell Riedel-de Haën™

Product Code. 15605740
Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
Aluminum Bottle
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

boiling range 93.5 - 100.4 °C

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-82-5
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098

Specifications

Melting Point -90.5°C
Molecular Formula C6H12
Linear Formula CH3CON(CH3)2
MDL Number MFCD00003814
UN Number UN1206
Beilstein 1730763
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
SMILES C1CCCCC1
IUPAC Name cyclohexane
Molecular Weight (g/mol) 84.16
Grade Performance
Density 0.71 g/mL at 20 °C
Boiling Point 83°C to 105°C
Quantity 5 L
Flash Point −7°C
Packaging Aluminum Bottle
Refractive Index 1.397
Chemical Name or Material Heptane Fraction
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'