missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptane Fraction, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
Aluminum Bottle
Glass Bottle
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Specifications
Melting Point | -90.5°C |
Molecular Formula | C7H16 |
MDL Number | MFCD00009544 |
UN Number | UN1206 |
Beilstein | 1730763 |
InChI Key | IMNFDUFMRHMDMM-UHFFFAOYSA-N |
SMILES | CCCCCCC |
IUPAC Name | heptane |
Molecular Weight (g/mol) | 100.21 |
Density | 0.71g/mL at 20°C |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'