missing translation for 'onlineSavingsMsg'
Learn More

Chloroiodomethane, 98%, stab. with copper, Thermo Scientific Chemicals

29.55 € - 323.61 €

Chemical Identifiers

CAS 593-71-5
Molecular Formula CH2ClI
MDL Number MFCD00001078
InChI Key PJGJQVRXEUVAFT-UHFFFAOYSA-N
Synonym chloro iodo methane, methane, chloroiodo, methylene chloroiodide, chloro-iodomethane, chloro-iodo-methane, iodochloromethane, chlor iod methan, chloroiodo-methane, chloromethyl iodide, qmablxaih@
PubChem CID 11644
IUPAC Name chloro(iodo)methane
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11384559
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
A11032.09
10 g
29.55 €
10g
Please to purchase this item. Need a web account? Register with us today!
11394559
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
A11032.18
50 g
65.02 €
50g
Please to purchase this item. Need a web account? Register with us today!
11304569
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
A11032.30
250 g
204.02 €
250g
Please to purchase this item. Need a web account? Register with us today!
11498863
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
A11032.36
500 g
323.61 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Chloroiodomethane is used as an intermediate in pharmaceuticals, organic synthesis, agrochemicals and dyestuff. It is also useful in cyclopropanation (Simmon-Smith reaction), Mannich reaction, aminomethylation and in epoxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroiodomethane is used as an intermediate in pharmaceuticals, organic synthesis, agrochemicals and dyestuff. It is also useful in cyclopropanation (Simmon-Smith reaction), Mannich reaction, aminomethylation and in epoxidation.

Solubility
Miscible with acetone, benzene, diethyl ether and alcohol.

Notes
It is incompatible with strong oxidizing agents and strong bases.
Specifications

Chemical Identifiers

593-71-5
MFCD00001078
chloro iodo methane, methane, chloroiodo, methylene chloroiodide, chloro-iodomethane, chloro-iodo-methane, iodochloromethane, chlor iod methan, chloroiodo-methane, chloromethyl iodide, qmablxaih@
chloro(iodo)methane
CH2ClI
PJGJQVRXEUVAFT-UHFFFAOYSA-N
11644

Specifications

593-71-5
108°C to 109°C
CH2ClI
MFCD00001078
1730802
chloro iodo methane, methane, chloroiodo, methylene chloroiodide, chloro-iodomethane, chloro-iodo-methane, iodochloromethane, chlor iod methan, chloroiodo-methane, chloromethyl iodide, qmablxaih@
PJGJQVRXEUVAFT-UHFFFAOYSA-N
chloro(iodo)methane
11644
98%
2.416
None
1.5822
UN2810
Light sensitive
Miscible with acetone,benzene,diethyl ether and alcohol.
C(Cl)I
176.381
176.38
Chloroiodomethane, Stabilized with copper
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 209-804-8

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.