missing translation for 'onlineSavingsMsg'
Learn More

Chloroiodomethane, 98%, stab. with copper, Thermo Scientific Chemicals

Product Code. 11304569
Click to view available options
Quantity:
10 g
50 g
250 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chloroiodomethane is used as an intermediate in pharmaceuticals, organic synthesis, agrochemicals and dyestuff. It is also useful in cyclopropanation (Simmon-Smith reaction), Mannich reaction, aminomethylation and in epoxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroiodomethane is used as an intermediate in pharmaceuticals, organic synthesis, agrochemicals and dyestuff. It is also useful in cyclopropanation (Simmon-Smith reaction), Mannich reaction, aminomethylation and in epoxidation.

Solubility
Miscible with acetone, benzene, diethyl ether and alcohol.

Notes
It is incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-71-5
Molecular Formula CH2ClI
MDL Number MFCD00001078
InChI Key PJGJQVRXEUVAFT-UHFFFAOYSA-N
Synonym chloro iodo methane, methane, chloroiodo, methylene chloroiodide, chloro-iodomethane, chloro-iodo-methane, iodochloromethane, chlor iod methan, chloroiodo-methane, chloromethyl iodide, qmablxaih@
PubChem CID 11644
IUPAC Name chloro(iodo)methane

Specifications

Density 2.416
Boiling Point 108°C to 109°C
Flash Point None
Refractive Index 1.5822
Quantity 250 g
UN Number UN2810
Beilstein 1730802
Sensitivity Light sensitive
Solubility Information Miscible with acetone,benzene,diethyl ether and alcohol.
SMILES C(Cl)I
Molecular Weight (g/mol) 176.381
Formula Weight 176.38
Percent Purity 98%
Chemical Name or Material Chloroiodomethane, Stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.