missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Sulfate, ≥99%, ACS reagent, Honeywell Fluka™
ACS Reagent,≥99.0%
39.69 € - 746.36 €
Chemical Identifiers
CAS | 7783-20-2 |
---|---|
Molecular Formula | H8N2O4S |
Molecular Weight (g/mol) | 132.13 |
MDL Number | MFCD00003391 |
InChI Key | BFNBIHQBYMNNAN-UHFFFAOYSA-N |
Synonym | ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate |
PubChem CID | 6097028 |
ChEBI | CHEBI:62946 |
SMILES | N.N.OS(O)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15697640
|
Honeywell-Fluka
A4915-25G |
25 g | |||||||
15617650
|
Honeywell-Fluka
A4915-500G |
500 g | |||||||
15677640
|
Honeywell-Fluka
A4915-1KG |
1 kg | |||||||
15687640
|
Honeywell-Fluka
A4915-2.5KG |
2.5 kg | |||||||
15637650
|
Honeywell-Fluka
A4915-6X500G |
6 x 500 g | |||||||
15627650
|
Honeywell-Fluka
A4915-5KG |
5 kg | |||||||
15667640
|
Honeywell-Fluka
A4915-12KG |
12 kg | |||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7783-20-2 | |
132.13 | |
BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
6097028 | |
N.N.OS(O)(=O)=O |
H8N2O4S | |
MFCD00003391 | |
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
CHEBI:62946 |
Specifications
7783-20-2 | |
(NH4)2SO4 | |
NONH for all modes of transport | |
BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
sulfuric acid diamine | |
6097028 | |
132.14g/mol | |
1.77g/mL at 25°C |
H8N2O4S | |
MFCD00003391 | |
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
N.N.OS(O)(=O)=O | |
132.13 | |
CHEBI:62946 | |
≥99% | |
Ammonium sulfate |
Safety and Handling
missing translation for 'rtecsNumber' : BS4500000