missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, ≥99%, ACS reagent, Honeywell Fluka™

Product Code. 15617650
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
5 kg
6 x 500 g
12 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
SMILES N.N.OS(O)(=O)=O

Specifications

Quantity 500 g
Linear Formula (NH4)2SO4
UN Number NONH for all modes of transport
IUPAC Name diazanium;sulfate
Formula Weight 132.14g/mol
Percent Purity ≥99%
Density 1.77g/mL at 25°C
Chemical Name or Material Ammonium sulfate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'