missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Sulfate, ≥99%, ACS reagent, Honeywell Fluka™
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
1 kg
2.5 kg
5 kg
6 x 500 g
12 kg
Beschreibung
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Spezifikation
Spezifikation
Quantity | 2.5 kg |
Linear Formula | (NH4)2SO4 |
UN Number | NONH for all modes of transport |
IUPAC Name | diazanium;sulfate |
Formula Weight | 132.14g/mol |
Percent Purity | ≥99% |
Density | 1.77g/mL at 25°C |
Chemical Name or Material | Ammonium sulfate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'