missing translation for 'onlineSavingsMsg'
Learn More

Ammonium acetate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥98%, Honeywell Fluka™

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥98%

41.96 € - 1291.17 €

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
MDL Number MFCD00013066
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]
View More Specs

Products 9
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15663780
View Documents Promotion Details
Honeywell-Fluka
32301-100G
100 g
41.96 €
100g
Please to purchase this item. Need a web account? Register with us today!
15613790
View Documents Promotion Details
Honeywell-Fluka
32301-500G
500 g
78.21 €
500g
Please to purchase this item. Need a web account? Register with us today!
15683780
View Documents Promotion Details
Honeywell-Fluka
32301-1KG
1 kg
132.50 €
1kg
Please to purchase this item. Need a web account? Register with us today!
15693780
View Documents Promotion Details
Honeywell-Fluka
32301-2.5KG
2.5 kg
256.86 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
15643790
View Documents Promotion Details
Honeywell-Fluka
32301-6X500G
6 x 500 g
371.87 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15623790
View Documents Promotion Details
Honeywell-Fluka
32301-6X1KG
6 x 1 kg
693.12 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15673780
View Documents Promotion Details
Honeywell-Fluka
32301-10KG
10 kg
736.33 €
10kg
Please to purchase this item. Need a web account? Register with us today!
15633790
View Documents Promotion Details
Honeywell-Fluka
32301-6X2.5KG
6 x 2.5 kg
1291.17 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15603790
View Documents Promotion Details
Honeywell-Fluka
32301-20KG
20 kg
1060.71 €
20kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

631-61-8
77.083
USFZMSVCRYTOJT-UHFFFAOYSA-N
517165
azanium;acetate
C2H7NO2
MFCD00013066
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
CHEBI:62947
CC(=O)[O-].[NH4+]

Specifications

631-61-8
CH3CO2NH4
4186741
USFZMSVCRYTOJT-UHFFFAOYSA-N
azanium;acetate
517165
77.08g/mol
ACS/puriss. p.a.
Ammonium Acetate, Reag. Ph. Eur.
C2H7NO2
MFCD00013066
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
CC(=O)[O-].[NH4+]
77.083
CHEBI:62947
≥98%
≤0.01% (as SO4)
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 211-162-9

missing translation for 'rtecsNumber' : AF3675000

Hazard Statement
  • EUH210-Safety data sheet available on request.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

SDS
Documents

Documents

Product Certifications