missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium acetate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥98%, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
6 x 500 g
10 kg
20 kg
6 x 1 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Melting Point | 110°C to 112°C (decomposition)(literature) |
CAS Max % | ≤100.0000% |
Quantity | 6 x 500 g |
Linear Formula | CH3CO2NH4 |
Beilstein | 4186741 |
Formula Weight | 77.08 |
Percent Purity | >98% |
Grade | ACS Reagent/Reagent Ph. Eur. |
Packaging | Poly Bottle |
Density | 1.170 g/cm3 |
Show More |
Safety and Handling
Hazard Statement
- EUH210-Safety data sheet available on request.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'