missing translation for 'onlineSavingsMsg'
Learn More

Ammonium acetate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥98%, Honeywell Fluka™

Product Code. 15633790
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
6 x 500 g
10 kg
20 kg
6 x 1 kg
6 x 2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
MDL Number MFCD00013066
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Specifications

Quantity 6 x 2.5 kg
Linear Formula CH3CO2NH4
Formula Weight 77.08
Percent Purity ≥98%
Grade ACS/puriss. p.a.
Residue after Ignition ≤0.01% (as SO4)
Chemical Name or Material Ammonium Acetate, Reag. Ph. Eur.
Hazard Statement
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'