Learn More
Ansell™ AlphaTec™ 57-435 Series Nitrile Immersion Gloves
Provides confident grip performance with enhanced flexibility. Ansell Alphatec™ 58-435 Series Nitrile Immersion Gloves has optimal control when handling oily or wet items.
119.00 €
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15599441
|
Ansell™
58435070 |
7 |
119.00 €
12 pairs |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
15509451
|
Ansell™
58435080 |
8 |
119.00 €
12 pairs |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
15519451
|
Ansell™
58435090 |
9 |
119.00 €
12 pairs |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
15385181
|
Ansell™
58435100 |
10 |
119.00 €
12 pairs |
In Stock
Log in to see stock. |
|||||
15375191
|
Ansell™
58435110 |
11 |
119.00 €
12 pairs |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
Description
- Chemical and liquid protection gloves
- Features Ansell Grip Technology™ for optimal control when handling oily or wet items
- Less force is required on slippery objects, reducing hand and arm fatigue
- Offers improved flexibility and dexterity thanks to cotton-flocked lined nitrile shell and snug, second-skin tailoring that tightens the fit across the palm
- Has 6-mil shell effective against hydrocarbon derivatives, alcohol bases, many solvents and esters
- Offers superior snag, puncture and abrasion protection for long-term wear
- Foldable gutter cuff helps prevent dripping onto the forearm, while an extra-long cuff extends protection to the upper forearm
Applications:
- Chemical handling
- Petrochemicals
- Rubber and rubber additives
- Handling fluid-coated metal parts
- Industrial tool maintenance
- Handling waste oils
- Waste and water treatment
Specifications
Gauntlet |