Learn More
Ansell™ AlphaTec™ 57-435 Series Nitrile Immersion Gloves
Product Code. 15375191
Click to view available options
Size:
7
8
9
10
11
Provides confident grip performance with enhanced flexibility. Ansell Alphatec™ 58-435 Series Nitrile Immersion Gloves has optimal control when handling oily or wet items.
  • Chemical and liquid protection gloves
  • Features Ansell Grip Technology™ for optimal control when handling oily or wet items
  • Less force is required on slippery objects, reducing hand and arm fatigue
  • Offers improved flexibility and dexterity thanks to cotton-flocked lined nitrile shell and snug, second-skin tailoring that tightens the fit across the palm
  • Has 6-mil shell effective against hydrocarbon derivatives, alcohol bases, many solvents and esters
  • Offers superior snag, puncture and abrasion protection for long-term wear
  • Foldable gutter cuff helps prevent dripping onto the forearm, while an extra-long cuff extends protection to the upper forearm
Applications:
  • Chemical handling
  • Petrochemicals
  • Rubber and rubber additives
  • Handling fluid-coated metal parts
  • Industrial tool maintenance
  • Handling waste oils
  • Waste and water treatment
TRUSTED_SUSTAINABILITY
Specifications
Back Material Nitrile
Cuff Material Nitrile
Product Type Immersion Gloves
Size 11
Style 57-435
Glove Cuff Style Gauntlet
Liner Material Cotton Flocking
Quantity 12 Pairs
Resistant To Chemical resistance
Series 57-435