missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ AlphaTec™ 57-435 Series Nitrile Immersion Gloves

Product Code. 15519451
Click to view available options
Size:
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Provides confident grip performance with enhanced flexibility. Ansell Alphatec™ 58-435 Series Nitrile Immersion Gloves has optimal control when handling oily or wet items.

  • Chemical and liquid protection gloves
  • Features Ansell Grip Technology™ for optimal control when handling oily or wet items
  • Less force is required on slippery objects, reducing hand and arm fatigue
  • Offers improved flexibility and dexterity thanks to cotton-flocked lined nitrile shell and snug, second-skin tailoring that tightens the fit across the palm
  • Has 6-mil shell effective against hydrocarbon derivatives, alcohol bases, many solvents and esters
  • Offers superior snag, puncture and abrasion protection for long-term wear
  • Foldable gutter cuff helps prevent dripping onto the forearm, while an extra-long cuff extends protection to the upper forearm

Applications:

  • Chemical handling
  • Petrochemicals
  • Rubber and rubber additives
  • Handling fluid-coated metal parts
  • Industrial tool maintenance
  • Handling waste oils
  • Waste and water treatment
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 374, EN 388:2003 4001
Description Grip design: Ansell Grip Technology™
Color Green, Gray
Latex Free Yes
Material Nitrile
Product Type Chemical and Liquid Protection Gloves
Size 9
Style Construction: Supported
Glove Cuff Style Gauntlet
Liner Material Cotton flocking
Packaging Type 12 pairs/polybag, 6 polybags/carton, 72 pairs/carton
Series 58-435
Length (Metric) 380 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'