Learn More
5alpha-Cholestan-3-one, 97%, Thermo Scientific Chemicals
61.20 € - 227.13 €
Chemical Identifiers
CAS | 566-88-1 |
---|---|
Molecular Formula | C27H46O |
Molecular Weight (g/mol) | 386.664 |
MDL Number | MFCD00065901 |
InChI Key | PESKGJQREUXSRR-UXIWKSIVSA-N |
Synonym | 5alpha-cholestan-3-one, 5a-cholestan-3-one, 5alpha-cholestanone, coprostanone, 5alpha-cholestane-3-one, cholestan-3-one, 5a, 3-keto-5alpha-cholestane, 5-alpha-cholestan-3-one, cholestan-3-one |
PubChem CID | 92128 |
ChEBI | CHEBI:17762 |
IUPAC Name | (5S,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-3-one |
SMILES | CC(C)CCCC(C)C1CCC2C1(CCC3C2CCC4C3(CCC(=O)C4)C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11362676
|
Thermo Scientific Alfa Aesar
L08726.03 |
1 g |
61.20 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11372676
|
Thermo Scientific Alfa Aesar
L08726.06 |
5 g |
227.13 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is employed as a intermediate for organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a intermediate for organic synthesis.
Solubility
Insoluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
566-88-1 | |
386.664 | |
PESKGJQREUXSRR-UXIWKSIVSA-N | |
92128 | |
(5S,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-3-one |
C27H46O | |
MFCD00065901 | |
5alpha-cholestan-3-one, 5a-cholestan-3-one, 5alpha-cholestanone, coprostanone, 5alpha-cholestane-3-one, cholestan-3-one, 5a, 3-keto-5alpha-cholestane, 5-alpha-cholestan-3-one, cholestan-3-one | |
CHEBI:17762 | |
CC(C)CCCC(C)C1CCC2C1(CCC3C2CCC4C3(CCC(=O)C4)C)C |
Specifications
566-88-1 | |
C27H46O | |
2625580 | |
Insoluble in water. | |
+43° (c=1 in Chloroform) | |
(5S,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-3-one | |
92128 | |
386.66 | |
5alpha-Cholestan-3-one |
128°C to 132°C | |
MFCD00065901 | |
5alpha-cholestan-3-one, 5a-cholestan-3-one, 5alpha-cholestanone, coprostanone, 5alpha-cholestane-3-one, cholestan-3-one, 5a, 3-keto-5alpha-cholestane, 5-alpha-cholestan-3-one, cholestan-3-one | |
PESKGJQREUXSRR-UXIWKSIVSA-N | |
CC(C)CCCC(C)C1CCC2C1(CCC3C2CCC4C3(CCC(=O)C4)C)C | |
386.664 | |
CHEBI:17762 | |
97% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.