missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5alpha-Cholestan-3-one, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
It is employed as a intermediate for organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a intermediate for organic synthesis.
Solubility
Insoluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
It is employed as a intermediate for organic synthesis.
Solubility
Insoluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Specifications
Melting Point | 128°C to 132°C |
Quantity | 5 g |
Beilstein | 2625580 |
Solubility Information | Insoluble in water. |
Optical Rotation | +43° (c=1 in Chloroform) |
Formula Weight | 386.66 |
Percent Purity | 97% |
Chemical Name or Material | 5alpha-Cholestan-3-one |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction