missing translation for 'onlineSavingsMsg'
Learn More

5alpha-Cholestan-3-one, 97%, Thermo Scientific Chemicals

Product Code. 11362676
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is employed as a intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 566-88-1
Molecular Formula C27H46O
Molecular Weight (g/mol) 386.664
MDL Number MFCD00065901
InChI Key PESKGJQREUXSRR-UXIWKSIVSA-N
Synonym 5alpha-cholestan-3-one, 5a-cholestan-3-one, 5alpha-cholestanone, coprostanone, 5alpha-cholestane-3-one, cholestan-3-one, 5a, 3-keto-5alpha-cholestane, 5-alpha-cholestan-3-one, cholestan-3-one
PubChem CID 92128
ChEBI CHEBI:17762
IUPAC Name (5S,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-3-one
SMILES CC(C)CCCC(C)C1CCC2C1(CCC3C2CCC4C3(CCC(=O)C4)C)C

Specifications

Melting Point 128°C to 132°C
Quantity 1 g
Beilstein 2625580
Solubility Information Insoluble in water.
Optical Rotation +43° (c=1 in Chloroform)
Formula Weight 386.66
Percent Purity 97%
Chemical Name or Material 5alpha-Cholestan-3-one

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.