missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dihydro-2H-pyran-4-yl trifluoromethanesulfonate, 95%, Thermo Scientific™

148.00 € - 1035.00 €

Chemical Identifiers

CAS 188975-30-6
Molecular Formula C6H7F3O4S
Molecular Weight (g/mol) 232.173
InChI Key QXCAHEXKUMUTRF-UHFFFAOYSA-N
Synonym 3,6-dihydro-2h-pyran-4-yl trifluoro-methanesulfonate, 5,6-dihydro-4-trifluoromethylsulfonyloxy-2h-pyran, 3,6-dihydro-2h-pyran-4-yltrifluoromethanesulfonate, 2,3-dihydro-4-trifluoromethylsulfonyloxy-6h-pyran, 2h-5,6-dihydropyran-4-yl trifluoromethyl sulfonate, 4-trifluoromethylsulfonyloxy-5,6-dihydro-2h-pyran, trifluoro-methanesulfonic acid 3,6-dihydro-2h-pyran-4-yl ester, trifluoromethanesulfonic acid 3,6-dihydro-2h-pyran-4-yl ester, methanesulfonic acid, 1,1,1-trifluoro-, 3,6-dihydro-2h-pyran-4-yl ester
PubChem CID 11356703
IUPAC Name 3,6-dihydro-2H-pyran-4-yl trifluoromethanesulfonate
SMILES C1COCC=C1OS(=O)(=O)C(F)(F)F
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10743512
View Documents Promotion Details
Thermo Scientific Maybridge
MO08368DA
1 g Amber Glass Bottle
148.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
10311610
View Documents Promotion Details
Thermo Scientific Maybridge
MO08368DE
5 g Amber Glass Bottle
590.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
10505470
View Documents Promotion Details
Thermo Scientific Maybridge
MO08368EA
10 g Amber Glass Bottle
1035.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

188975-30-6
232.173
3,6-dihydro-2h-pyran-4-yl trifluoro-methanesulfonate, 5,6-dihydro-4-trifluoromethylsulfonyloxy-2h-pyran, 3,6-dihydro-2h-pyran-4-yltrifluoromethanesulfonate, 2,3-dihydro-4-trifluoromethylsulfonyloxy-6h-pyran, 2h-5,6-dihydropyran-4-yl trifluoromethyl sulfonate, 4-trifluoromethylsulfonyloxy-5,6-dihydro-2h-pyran, trifluoro-methanesulfonic acid 3,6-dihydro-2h-pyran-4-yl ester, trifluoromethanesulfonic acid 3,6-dihydro-2h-pyran-4-yl ester, methanesulfonic acid, 1,1,1-trifluoro-, 3,6-dihydro-2h-pyran-4-yl ester
3,6-dihydro-2H-pyran-4-yl trifluoromethanesulfonate
C6H7F3O4S
QXCAHEXKUMUTRF-UHFFFAOYSA-N
11356703
C1COCC=C1OS(=O)(=O)C(F)(F)F

Specifications

188975-30-6
C6H7F3O4S
QXCAHEXKUMUTRF-UHFFFAOYSA-N
3,6-dihydro-2H-pyran-4-yl trifluoromethanesulfonate
11356703
3, 6-Dihydro-2H-pyran-4-yl trifluoromethanesulfonate
95%
3,6-dihydro-2h-pyran-4-yl trifluoro-methanesulfonate, 5,6-dihydro-4-trifluoromethylsulfonyloxy-2h-pyran, 3,6-dihydro-2h-pyran-4-yltrifluoromethanesulfonate, 2,3-dihydro-4-trifluoromethylsulfonyloxy-6h-pyran, 2h-5,6-dihydropyran-4-yl trifluoromethyl sulfonate, 4-trifluoromethylsulfonyloxy-5,6-dihydro-2h-pyran, trifluoro-methanesulfonic acid 3,6-dihydro-2h-pyran-4-yl ester, trifluoromethanesulfonic acid 3,6-dihydro-2h-pyran-4-yl ester, methanesulfonic acid, 1,1,1-trifluoro-, 3,6-dihydro-2h-pyran-4-yl ester
C1COCC=C1OS(=O)(=O)C(F)(F)F
232.173
232.18
Videos
Safety and Handling

Safety and Handling

Warning

SDS
Documents

Documents

Product Certifications