missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Dihydro-2H-pyran-4-yl trifluoromethanesulfonate, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 232.18 |
Chemical Name or Material | 3, 6-Dihydro-2H-pyran-4-yl trifluoromethanesulfonate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'