missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dihydro-2H-pyran-4-yl trifluoromethanesulfonate, 95%, Thermo Scientific™

Product Code. 10505470
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 188975-30-6
Molecular Formula C6H7F3O4S
Molecular Weight (g/mol) 232.173
InChI Key QXCAHEXKUMUTRF-UHFFFAOYSA-N
Synonym 3,6-dihydro-2h-pyran-4-yl trifluoro-methanesulfonate, 5,6-dihydro-4-trifluoromethylsulfonyloxy-2h-pyran, 3,6-dihydro-2h-pyran-4-yltrifluoromethanesulfonate, 2,3-dihydro-4-trifluoromethylsulfonyloxy-6h-pyran, 2h-5,6-dihydropyran-4-yl trifluoromethyl sulfonate, 4-trifluoromethylsulfonyloxy-5,6-dihydro-2h-pyran, trifluoro-methanesulfonic acid 3,6-dihydro-2h-pyran-4-yl ester, trifluoromethanesulfonic acid 3,6-dihydro-2h-pyran-4-yl ester, methanesulfonic acid, 1,1,1-trifluoro-, 3,6-dihydro-2h-pyran-4-yl ester
PubChem CID 11356703
IUPAC Name 3,6-dihydro-2H-pyran-4-yl trifluoromethanesulfonate
SMILES C1COCC=C1OS(=O)(=O)C(F)(F)F

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 232.18
Chemical Name or Material 3, 6-Dihydro-2H-pyran-4-yl trifluoromethanesulfonate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'