missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethoxyethanol, Reagent Grade, 99%, Honeywell™
Reagent Grade, 99%
28.92 € - 184.06 €
Chemical Identifiers
CAS | 110-80-5 |
---|---|
Molecular Formula | C4H10O2 |
Molecular Weight (g/mol) | 90.12 |
MDL Number | MFCD00002869 |
InChI Key | ZNQVEEAIQZEUHB-UHFFFAOYSA-N |
Synonym | cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee |
PubChem CID | 8076 |
ChEBI | CHEBI:46788 |
SMILES | CCOCCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15621200
|
Honeywell Research Chemicals
128082-500ML |
500 mL | Glass Bottle | ||||||
15691190
|
Honeywell Research Chemicals
128082-1L |
1 L | Glass Bottle |
68.73 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15601200
|
Honeywell Research Chemicals
128082-2.5L |
2.5 L | Glass Bottle |
184.06 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
110-80-5 | |
90.12 | |
ZNQVEEAIQZEUHB-UHFFFAOYSA-N | |
8076 | |
CCOCCO |
C4H10O2 | |
MFCD00002869 | |
cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee | |
CHEBI:46788 |
Specifications
110-80-5 | |
-90°C (literature) | |
136°C | |
C4H10O2 | |
MFCD00002869 | |
1098271 | |
ZNQVEEAIQZEUHB-UHFFFAOYSA-N | |
2-ethoxyethan-1-ol | |
8076 | |
90.12 | |
Reagent |
100.0000% | |
0.93 g/mL at 25 °C(lit.) | |
40°C | |
FeCl3 | |
UN1171 | |
cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee | |
CCOCCO | |
90.12 | |
CHEBI:46788 | |
>98.5% | |
2-Ethoxyethanol, Reagent Grade, 99% |
Safety and Handling
missing translation for 'shelfLife' : 1080 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Ethylene glycol monoethyl ether
missing translation for 'einecsNumber' : 203-804-1
Recommended Storage : Room Temp