missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethoxyethanol, Reagent Grade, 99%, Honeywell™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
CAS Max % | 100.0000% |
Melting Point | -90°C (literature) |
Density | 0.93 g/mL at 25 °C(lit.) |
Boiling Point | 136°C |
Flash Point | 40°C |
Packaging | Glass Bottle |
Linear Formula | FeCl3 |
Quantity | 2.5 L |
UN Number | UN1171 |
Beilstein | 1098271 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'