missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyethanol, Reagent Grade, 99%, Honeywell™

Product Code. 15621200
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-80-5
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00002869
InChI Key ZNQVEEAIQZEUHB-UHFFFAOYSA-N
Synonym cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee
PubChem CID 8076
ChEBI CHEBI:46788
SMILES CCOCCO

Specifications

CAS Max % 100.0000%
Melting Point -90°C (literature)
Density 0.93 g/mL at 25 °C(lit.)
Boiling Point 136°C
Flash Point 40°C
Packaging Glass Bottle
Linear Formula FeCl3
Quantity 500 mL
UN Number UN1171
Beilstein 1098271
IUPAC Name 2-ethoxyethan-1-ol
Formula Weight 90.12
Percent Purity >98.5%
Grade Reagent
Chemical Name or Material 2-Ethoxyethanol, Reagent Grade, 99%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'