missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Amber Boston Rounds with Polyvinyl-lined Caps
Amber Type III soda lime glass protects light-sensitive solutions
31.45 € - 355.00 €
Specifications
Color | Amber |
---|---|
Material | Soda Lime Glass |
Closure Color | White |
Closure Liner | Poly/Vinyl |
Closure Material | Polypropylene |
Product Code | Brand | Capacity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Packaging | Price | Quantity & Availability | ||||
10396992
|
DWK Life Sciences
W216836 |
1 oz., 30mL | Convenience pack |
66.25 €
Pack of 48 |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||
10691993
|
DWK Life Sciences
W216837 |
2 oz., 60 mL | Convenience pack |
42.40 €
Pack of 24 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
12339779
|
DWK Life Sciences
W216860 |
2 oz., 60 mL | Valu-bulk pack |
355.00 €
Pack of 288 |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||
12349779
|
DWK Life Sciences
W216861 |
4 oz., 125 mL | Valu-bulk Pk. |
260.00 €
Pack of 160 |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||
10541043
|
DWK Life Sciences
W216838 |
4 oz., 125 mL | Convenience pack |
44.10 €
Pack of 24 |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||
10708524
|
DWK Life Sciences
W216839 |
8 oz., 250 mL | Convenience pack |
31.45 €
Pack of 12 |
Estimated Shipment: 15-04-2025
Log in to see stock. |
||||
10225262
|
DWK Life Sciences
W216840 |
16 oz., 500 mL | Convenience pack |
37.90 €
Pack of 12 |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||
12369779
|
DWK Life Sciences
W216863 |
16 oz., 500 mL | Valu-bulk pack |
159.00 €
Pack of 60 |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||
12379779
|
DWK Life Sciences
W216864 |
32 oz., 1000 mL | Valu-bulk pack |
122.00 €
Pack of 30 |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||
10011362
|
DWK Life Sciences
W216841 |
32 oz., 1000 mL | Convenience pack |
47.45 €
Pack of 12 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
Description
Amber Type III soda lime glass protects light-sensitive solutions
- Available in partitioned Convenience Pack or Valu-Bulk™ cartons
- Convenience Pack bottles come with Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage
Each case of Convenience Pack bottles includes Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage
Order Info
Caps can be ordered separately.
Compliance
USP Type III soda-lime glass
Specifications
Amber | |
White | |
Polypropylene |
Soda Lime Glass | |
Poly/Vinyl | |
Screw Cap |